missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Camphor, Synthetic, USP, Spectrum™ Chemical
CA185, 76-22-2, C10H16O
Supplier: Spectrum Chemical Mfg Cor CA18525KGBL
Description
Spectrum™ Chemical Camphor, Synthetic, USP is used as a decongestant, antiseptic stimulant and anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 76-22-2 | |
| C10H16O | |
| UN2717 | |
| DSSYKIVIOFKYAU-UHFFFAOYNA-N | |
| 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one | |
| USP |
| 100% | |
| MFCD00074738,MFCD00064149 | |
| 25 kg | |
| CC1(C)C2CCC1(C)C(=O)C2 | |
| 152.24 | |
| Fiber Drum |
Chemical Identifiers
| 76-22-2 | |
| 152.24 | |
| DSSYKIVIOFKYAU-UHFFFAOYNA-N | |
| CC1(C)C2CCC1(C)C(=O)C2 |
| C10H16O | |
| MFCD00074738,MFCD00064149 | |
| 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one |