missing translation for 'onlineSavingsMsg'
Learn More

Camphor, Synthetic, USP, Spectrum™ Chemical

Catalog No. p-7157246
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
110 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601403 500 g Amber Glass Bottle
18601407 110 lb. Fiber Drum
18601404 2.5 kg Poly Pail
18601405 12 kg Fiber Drum
18601406 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601403 Supplier Spectrum Chemical Mfg Cor Supplier No. CA185500GM
Only null left

CA185, 76-22-2, C10H16O

Spectrum™ Chemical Camphor, Synthetic, USP is used as a decongestant, antiseptic stimulant and anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 76-22-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074738,MFCD00064149
InChI Key DSSYKIVIOFKYAU-UHFFFAOYNA-N
IUPAC Name 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
SMILES CC1(C)C2CCC1(C)C(=O)C2

Specifications

CAS Max % 100%
UN Number UN2717
Quantity 500 g
Grade USP
Packaging Amber Glass Bottle