missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ cAMP-Screen™ Assay/Lysis Buffer

Catalog No. T2327
Encompass
Change view
Click to view available options
Quantity:
65 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2327 65 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2327 Supplier Invitrogen™ Supplier No. T2327

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with the cAMP-Screen or cAmp-Screen Direct™ Immunoassay System.

This Assay/Lysis Buffer is suitable for use with the cAMP-Screen™ or cAmp-Screen Direct™ Immunoassay System.

For Research Use Only. Not for use in diagnostics procedures.

  • Quantity: 65 mL

Specifications

Product Type ELISA Assay/Lysis Buffer
Quantity 65 mL
Form Liquid
For Use With (Equipment) cAmp-Screen Direct™ Immunoassay System., cAMP-Screen™ Immunoassay System
Product Line cAMP-Screen
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.