missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Camostat Mesylate 98.0+%, TCI America™
Supplier: TCI America C2977100MG
Specifications
| Camostat Mesylate | |
| 165°C | |
| C21H26N4O8S | |
| 100 mg | |
| FSEKIHNIDBATFG-UHFFFAOYSA-N | |
| 4-{2-[(dimethylcarbamoyl)methoxy]-2-oxoethyl}phenyl 4-[(diaminomethylidene)amino]benzoate; methanesulfonic acid | |
| 5284360 | |
| ≥98.0% (HPLC) |
| 59721-29-8 | |
| White | |
| MFCD00941410 | |
| camostat mesylate, camostat mesilate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238 | |
| CS(O)(=O)=O.CN(C)C(=O)COC(=O)CC1=CC=C(OC(=O)C2=CC=C(C=C2)N=C(N)N)C=C1 | |
| 494.52 | |
| 494.52 | |
| Crystalline Powder |
Chemical Identifiers
| 59721-29-8 | |
| 494.52 | |
| FSEKIHNIDBATFG-UHFFFAOYSA-N | |
| 5284360 | |
| CS(O)(=O)=O.CN(C)C(=O)COC(=O)CC1=CC=C(OC(=O)C2=CC=C(C=C2)N=C(N)N)C=C1 |
| C21H26N4O8S | |
| MFCD00941410 | |
| camostat mesylate, camostat mesilate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238 | |
| 4-{2-[(dimethylcarbamoyl)methoxy]-2-oxoethyl}phenyl 4-[(diaminomethylidene)amino]benzoate; methanesulfonic acid |
Safety and Handling
RTECSNumber : CY1571620
TSCA : No