missing translation for 'onlineSavingsMsg'
Learn More

Camostat Mesylate 98.0+%, TCI America™

Catalog No. p-7125756
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C297725MG 25 mg
C2977100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C297725MG Supplier TCI America Supplier No. C297725MG
Only null left

Chemical Identifiers

CAS 59721-29-8
Molecular Formula C21H26N4O8S
Molecular Weight (g/mol) 494.52
MDL Number MFCD00941410
InChI Key FSEKIHNIDBATFG-UHFFFAOYSA-N
Synonym camostat mesylate, camostat mesilate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238
PubChem CID 5284360
IUPAC Name 4-{2-[(dimethylcarbamoyl)methoxy]-2-oxoethyl}phenyl 4-[(diaminomethylidene)amino]benzoate; methanesulfonic acid
SMILES CS(O)(=O)=O.CN(C)C(=O)COC(=O)CC1=CC=C(OC(=O)C2=CC=C(C=C2)N=C(N)N)C=C1

Specifications

Melting Point 165°C
Color White
Quantity 25 mg
Formula Weight 494.52
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Camostat Mesylate
RTECSNumber CY1571620
TSCA No