missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calpain Inhibitor XI

$208.92 - $800.84

Chemical Identifiers

CAS 145731-49-3
Molecular Formula C26H40N4O6
Synonym Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65991MCR
View Documents Promotion Details
Thermo Scientific Chemicals
J65991MCR
1 mg
Each for $208.92
Only null left
 
AAJ65991LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J65991LB0
5 mg
Each for $800.84
Only null left
 
Description

Description

A cell-permeable dipeptidyl --ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

145731-49-3
C26H40N4O6
Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine

Specifications

Off-white
A cell-permeable dipeptidyl →-ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2
C26H40N4O6
Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine
Solid
CCC(NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C(=O)C(=O)NCCCN1CCOCC1
504.63
2°C to 8°C
145731-49-3
504.60
Light Sensitive
TZVQRMYLYQNBOA-UHFFFAOYNA-N
benzyl N-{3-methyl-1-[(1-{[3-(morpholin-4-yl)propyl]carbamoyl}-1-oxobutan-2-yl)carbamoyl]butyl}carbamate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only