missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Calpain Inhibitor XI
Description
A cell-permeable dipeptidyl --ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 mg |
| Description | Off-white |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | A cell-permeable dipeptidyl →-ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2 |
| Formula Weight | 504.60 |
| Sensitivity | Light Sensitive |
| Form | Solid |
RUO – Research Use Only