Learn More
Thermo Scientific Chemicals Calpain Inhibitor XI
Supplier:  Thermo Scientific Chemicals J65991LB0
Quantity 5 mg
 View more versions of this product
Catalog No. AAJ65991LB0
Only null left
Description
Description
A cell-permeable dipeptidyl --ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Identifiers
145731-49-3
C26H40N4O6
Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine
Specifications
5 mg
2°C to 8°C
145731-49-3
504.60
Light Sensitive
TZVQRMYLYQNBOA-UHFFFAOYNA-N
benzyl N-{3-methyl-1-[(1-{[3-(morpholin-4-yl)propyl]carbamoyl}-1-oxobutan-2-yl)carbamoyl]butyl}carbamate
Off-white
A cell-permeable dipeptidyl →-ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2
C26H40N4O6
Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine
Solid
CCC(NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C(=O)C(=O)NCCCN1CCOCC1
504.63
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Keep cold; Store under Nitrogen