missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate TS, Saturated, Ricca Chemical

Catalog No. 182354
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
120 mL
Packaging:
Natural Poly Bottle
Catalog No. Quantity Packaging
182354 120 mL Natural Poly Bottle
1 options

Catalog No. 182354

Supplier: Ricca Chemical Company 1823.54

Only null left

Chemical Identifiers

CAS 7732-18-5 , 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
CAS Min % 93.0, 5.49
CAS Max % 96.0, 5.71
Physical Form Liquid
Quantity 120 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) Saturated Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Calcium Sulfate TS
ShelfLife 12 months