missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium sulfate dihydrate, Puriss., meets analytical spec. of NF, E 516, 99.0-101.0% (based on anhydrous substance), Honeywell Fluka™
Puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
$293.73 - $293.73
Chemical Identifiers
| CAS | 10101-41-4 |
|---|---|
| Molecular Formula | CaH4O6S |
| Molecular Weight (g/mol) | 172.16 |
| MDL Number | MFCD00149625 |
| InChI Key | PASHVRUKOFIRIK-UHFFFAOYSA-L |
| Synonym | calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone |
| PubChem CID | 24928 |
| ChEBI | CHEBI:32583 |
| IUPAC Name | calcium;sulfate;dihydrate |
| SMILES | O.O.[Ca++].[O-]S([O-])(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 10101-41-4 | |
| 172.16 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| 24928 | |
| calcium;sulfate;dihydrate |
| CaH4O6S | |
| MFCD00149625 | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| CHEBI:32583 | |
| O.O.[Ca++].[O-]S([O-])(=O)=O |
Specifications
| 10101-41-4 | |
| CaH4O6S | |
| MFCD00149625 | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| O.O.[Ca++].[O-]S([O-])(=O)=O | |
| 172.16 | |
| CHEBI:32583 | |
| 99 to 101% (based on anhydrous substance) | |
| Calcium Sulfate Dihydrate, Meets analytical specification of NF |
| 1 kg | |
| CaSO4 · 2H2O | |
| NONH for all modes of transport | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| calcium;sulfate;dihydrate | |
| 24928 | |
| 172.17g/mol | |
| puriss. |
Safety and Handling
EINECSNumber : 231-900-3
RTECSNumber : EW4150000