missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, Dihydrate, FCC, 98%, Spectrum™ Chemical
CA173, 10101-41-4, CaSO4·2H2O
Supplier: Spectrum Chemical Mfg Cor CA17325KG
Description
Spectrum™ Chemical Calcium Sulfate, Dihydrate, FCC is widely used as pH regulator, hardness regulator, yeast agent, nutritional suppliment, texture and flavor enhancer in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 10101-41-4 | |
| CaH4O6S | |
| 2.5 kg | |
| O.O.[Ca++].[O-]S([O-])(=O)=O | |
| 172.16 | |
| FCC | |
| Poly Pail |
| 100% | |
| MFCD00149625 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| calcium dihydrate sulfate | |
| 98% | |
| 19.0-23% |
Chemical Identifiers
| 10101-41-4 | |
| 172.16 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| O.O.[Ca++].[O-]S([O-])(=O)=O |
| CaH4O6S | |
| MFCD00149625 | |
| calcium dihydrate sulfate |