missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate, Dihydrate, FCC, 98%, Spectrum™ Chemical

Catalog No. p-7157244
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18601395 500 g Poly Bottle
18601396 2.5 kg Poly Pail
18601397 12 kg Poly Pail
3 options

Catalog No. 18601395

Supplier: Spectrum Chemical Mfg Cor CA173500GM

Only null left

CA173, 10101-41-4, CaSO4·2H2O

Spectrum™ Chemical Calcium Sulfate, Dihydrate, FCC is widely used as pH regulator, hardness regulator, yeast agent, nutritional suppliment, texture and flavor enhancer in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98%
Grade FCC
Loss on Drying 19.0-23%
Packaging Poly Bottle