missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Supplier: MP Biomedicals Inc 0219141490
Specifications
| Calcium Sulfate Dihydrate | |
| Crystalline | |
| 98.0-102.0% | |
| MFCD00149625 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| calcium dihydrate sulfate | |
| 24928 | |
| 172.2 |
| 10101-41-4 | |
| 500 g | |
| CaH4O6S | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| O.O.[Ca++].[O-]S([O-])(=O)=O | |
| 172.16 | |
| CHEBI:32583 | |
| ACS Reagent |
Chemical Identifiers
| 10101-41-4 | |
| 172.16 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| 24928 | |
| calcium dihydrate sulfate |
| CaH4O6S | |
| MFCD00149625 | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| CHEBI:32583 | |
| O.O.[Ca++].[O-]S([O-])(=O)=O |
Safety and Handling
Recommended Storage : Room Temperature