missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Catalog No. p-3547049
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19141480 100 g
ICN19141490 500 g
ICN19141491 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19141480 Supplier MP Biomedicals Inc Supplier No. 0219141480
Only null left

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.0-102.0%
Formula Weight 172.2
Grade ACS Reagent
Chemical Name or Material Calcium Sulfate Dihydrate
Recommended Storage Room Temperature