missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Manufacturer: Mp Biomedicals 0219141491
Specifications
Calcium Sulfate Dihydrate | |
98.0-102.0% | |
annaline, calcium sulfate dihydrate, compactrol, gips, gypsite, gypsum stone, landplaster, phosphogypsum, primoplast, satinite | |
O.O.[O-]S(=O)(=O)[O-].[Ca+2] | |
172.164 | |
CHEBI:32583 | |
Crystalline | |
1kg |
10101-41-4 | |
CaH4O6S | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
calcium;sulfate;dihydrate | |
24928 | |
172.2 | |
ACS Reagent |
Chemical Identifiers
10101-41-4 | |
172.164 | |
annaline, calcium sulfate dihydrate, compactrol, gips, gypsite, gypsum stone, landplaster, phosphogypsum, primoplast, satinite | |
CHEBI:32583 | |
O.O.[O-]S(=O)(=O)[O-].[Ca+2] |
CaH4O6S | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
24928 | |
calcium;sulfate;dihydrate |
Safety and Handling
Recommended Storage : Room Temperature