missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Stearate, NF, Spectrum™ Chemical
CA169, 1592-23-0
Supplier: Spectrum Chemical Mfg Cor CA16912KG
Description
Spectrum™ Chemical Calcium Stearate, NF has good anti-adherent and lubricant properties and is used for tablet and capsule lubrication. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 1592-23-0 | |
| C36H70CaO4 | |
| CJZGTCYPCWQAJB-UHFFFAOYSA-L | |
| calcium dioctadecanoate | |
| NF | |
| Fiber Drum |
| 100% | |
| 12 kg | |
| [Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O | |
| 607.03 | |
| 4% |
Chemical Identifiers
| 1592-23-0 | |
| 607.03 | |
| calcium dioctadecanoate |
| C36H70CaO4 | |
| CJZGTCYPCWQAJB-UHFFFAOYSA-L | |
| [Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O |