missing translation for 'onlineSavingsMsg'
Learn More

Calcium Stearate, NF, Spectrum™ Chemical

Catalog No. p-7157241
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601385 500 g Poly Bottle
18601386 2.5 kg Poly Pail
18601387 12 kg Fiber Drum
18601388 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601385 Supplier Spectrum Chemical Mfg Cor Supplier No. CA169500GM
Only null left

CA169, 1592-23-0

Spectrum™ Chemical Calcium Stearate, NF has good anti-adherent and lubricant properties and is used for tablet and capsule lubrication. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1592-23-0
Molecular Formula C36H70CaO4
Molecular Weight (g/mol) 607.03
InChI Key CJZGTCYPCWQAJB-UHFFFAOYSA-L
IUPAC Name calcium dioctadecanoate
SMILES [Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Grade NF
Loss on Drying 4%
Packaging Poly Bottle