missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard, 1000 ppm Ca, Ricca Chemical

$286.80 - $641.77

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
1821-4
View Documents
Ricca Chemical Company
18214
120 mL Natural Poly Bottle
Each for $286.80
Only null left
 
1821-16
View Documents
Ricca Chemical Company
182116
500 mL Natural Poly Bottle
Each for $641.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
10112
calcium carbonate

Specifications

Natural Poly Bottle
1,000ppm Ca
100.0,0.38,0.25
CCaO3
Colorless
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
120 mL
CHEBI:3311
471-34-1,64-19-7,7732-18-5
97.0,0.37,0.24
Liquid
MFCD00010906
100°C
[Ca++].[O-]C([O-])=O
100.09
10112
Calcium Standard Solution, Stock Solution
Videos
SDS
Documents

Documents