missing translation for 'onlineSavingsMsg'
Learn More

Calcium propionate hydrate, 97%

CAS: 4075-81-4 | C6H10CaO4 | 186.22 g/mol

Supplier:  Thermo Scientific Chemicals A149940E

 View more versions of this product

Catalog No. AAA149940E


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium propionate hydrate
>300°C
MFCD00167354
Hygroscopic
calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907
[Ca++].CCC([O-])=O.CCC([O-])=O
186.22
CHEBI:81716
97%
4075-81-4
C6H10CaO4
2500 g
14,1698
BCZXFFBUYPCTSJ-UHFFFAOYSA-L
calcium;propanoate
19999
186.22 (Anhydrous)

Chemical Identifiers

4075-81-4
186.22
BCZXFFBUYPCTSJ-UHFFFAOYSA-L
19999
calcium;propanoate
C6H10CaO4
MFCD00167354
calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907
CHEBI:81716
[Ca++].CCC([O-])=O.CCC([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 223-795-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only