accessibility menu, dialog, popup

UserName

CaH3COOH3COO

CAS RN 4075-81-4

IUPAC Name: calcium dipropanoate
Synonyms: calcium propanoate|calcium propionate
Molecular Weight (g/mol): 186.22
Molecular Formula: C6H10CaO4
InChi Key: BCZXFFBUYPCTSJ-UHFFFAOYSA-L
SMILES: [Ca++].CCC([O-])=O.CCC([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (4)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (4)