missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate, Tribasic, Powder, FCC, 34-40%, Spectrum™ Chemical
C1144, 1306-06-5, Ca5OH(PO4)3
$689.57 - $9269.93
Chemical Identifiers
| CAS | 1306-06-5 |
|---|---|
| Molecular Formula | Ca5HO13P3 |
| Molecular Weight (g/mol) | 502.31 |
| MDL Number | MFCD00010904 |
| InChI Key | XYJRXVWERLGGKC-UHFFFAOYSA-D |
| IUPAC Name | pentacalcium hydroxide triphosphate |
| SMILES | [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601358
|
Spectrum Chemical Mfg Cor
C1144500GM |
500 g | Poly Bottle |
Each for $689.57
|
|
||||
|
18601359
|
Spectrum Chemical Mfg Cor
C114425KG |
2.5 kg | Poly Pail |
Each for $2,331.87
|
|
||||
|
18601360
|
Spectrum Chemical Mfg Cor
C114412KG |
12 kg | Fiber Drum |
Each for $4,887.60
|
|
||||
|
18601361
|
Spectrum Chemical Mfg Cor
C114445KGBL |
45 kg | Fiber Drum |
Each for $9,269.93
|
|
||||
Description
Spectrum™ Chemical Calcium Phosphate, Tribasic, Powder, FCC is used in powdered spices as an anticaking agent, a raising agent, an antacid and nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 1306-06-5 | |
| 502.31 | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
| Ca5HO13P3 | |
| MFCD00010904 | |
| pentacalcium hydroxide triphosphate |
Specifications
| 1306-06-5 | |
| 500 g | |
| MFCD00010904 | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
| 502.31 | |
| FCC | |
| Poly Bottle |
| 100% | |
| Ca5HO13P3 | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| pentacalcium hydroxide triphosphate | |
| 34 to 40% | |
| 10% |