missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Tribasic, Powder, FCC, 34-40%, Spectrum™ Chemical

Catalog No. 18601358
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601358 500 g Poly Bottle
18601359 2.5 kg Poly Pail
18601360 12 kg Fiber Drum
18601361 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601358 Supplier Spectrum Chemical Mfg Cor Supplier No. C1144500GM
Only null left

C1144, 1306-06-5, Ca5OH(PO4)3

Spectrum™ Chemical Calcium Phosphate, Tribasic, Powder, FCC is used in powdered spices as an anticaking agent, a raising agent, an antacid and nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1306-06-5
Molecular Formula Ca5HO13P3
Molecular Weight (g/mol) 502.31
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
IUPAC Name pentacalcium hydroxide triphosphate
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 34 to 40%
Grade FCC
Loss on Ignition 10%
Packaging Poly Bottle