missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate, Tribasic, Powder, FCC, 34-40%, Spectrum™ Chemical
Description
Spectrum™ Chemical Calcium Phosphate, Tribasic, Powder, FCC is used in powdered spices as an anticaking agent, a raising agent, an antacid and nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| Physical Form | Powder, Crystals and/or Chunks |
| CAS Max % | 100% |
| Quantity | 45 kg |
| Loss on Ignition | 10% |
| Packaging | Poly Bottle |
| Percent Purity | 34 to 40% |
| Grade | FCC |
| Chemical Name or Material | Calcium Phosphate, Tribasic |