missing translation for 'onlineSavingsMsg'
Learn More

Calcium Peroxide, FCC, 60%, Spectrum™ Chemical

C1388, 1305-79-9, CaO2

Supplier:  Spectrum Chemical Mfg Cor C1388500GM

 View more versions of this product

Catalog No. 18601343


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Calcium Peroxide, FCC is used as a food additive under the E number E930 as flour bleaching agent. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

1305-79-9
CaO2
500 g
[Ca++].[O-][O-]
72.08
FCC
100%
UN1457
LHJQIRIGXXHNLA-UHFFFAOYSA-N
calcium dioxidanediide
60%
Amber Glass Bottle

Chemical Identifiers

1305-79-9
72.08
calcium dioxidanediide
CaO2
LHJQIRIGXXHNLA-UHFFFAOYSA-N
[Ca++].[O-][O-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions