missing translation for 'onlineSavingsMsg'
Learn More

Calcium Peroxide, FCC, 60%, Spectrum™ Chemical

Catalog No. p-7157159
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
100 lb.
Packaging:
Amber Glass Bottle
Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601341 25 g Amber Glass Bottle
18601342 125 g Amber Glass Bottle
18601343 500 g Amber Glass Bottle
18601345 100 lb. Drum
18601344 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601341 Supplier Spectrum Chemical Mfg Cor Supplier No. C138825GM
Only null left

C1388, 1305-79-9, CaO2

Spectrum™ Chemical Calcium Peroxide, FCC is used as a food additive under the E number E930 as flour bleaching agent. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1305-79-9
Molecular Formula CaO2
Molecular Weight (g/mol) 72.08
InChI Key LHJQIRIGXXHNLA-UHFFFAOYSA-N
IUPAC Name calcium dioxidanediide
SMILES [Ca++].[O-][O-]

Specifications

CAS Max % 100%
UN Number UN1457
Quantity 25 g
Percent Purity 60%
Grade FCC
Packaging Amber Glass Bottle