accessibility menu, dialog, popup

UserName

CaOO

CAS RN 1305-79-9

IUPAC Name: calcium dioxidanediide
Synonyms: calcium peroxide
Molecular Weight (g/mol): 72.08
Molecular Formula: CaO2
InChi Key: LHJQIRIGXXHNLA-UHFFFAOYSA-N
SMILES: [Ca++].[O-][O-]
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)