missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide TS (Saturated Aqueous Solution), Ricca Chemical

Catalog No. 178532
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
178532 1 L Natural Poly Bottle
178516 500 mL Natural Poly Bottle
17851 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 178532 Supplier Ricca Chemical Company Supplier No. 178532

Chemical Identifiers

CAS 1305-62-0 , 7732-18-5
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Color Colorless
CAS Min % 98.0, 0.29
CAS Max % 100.0, 0.31
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Saturated Aqueous Solution
pH ∼12.4
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Calcium Hydroxide, Test Solution