missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Powder, EP, BP, JP, USP, 95-100.5%, Spectrum™ Chemical

CA150, 1305-62-0, Ca(OH)2

Supplier:  Spectrum Chemical Mfg Cor CA15012KG

 View more versions of this product

Catalog No. 18601322


Only null left

Description

Description

Spectrum™ Chemical Calcium Hydroxide, Powder, USP, EP, BP, JP is used as an astringent that is topically applied in solutions or lotions. Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Calcium Hydroxide
1305-62-0
CaH2O2
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium dihydroxide
95 to 100.5%
Poly Pail
White
100%
MFCD00010901
[OH-].[OH-].[Ca++]
74.09
Multi-Compendial
12 kg

Chemical Identifiers

1305-62-0
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
[OH-].[OH-].[Ca++]
CaH2O2
MFCD00010901
calcium dihydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions