missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Powder, EP, BP, JP, USP, 95-100.5%, Spectrum™ Chemical

Catalog No. p-7157234
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601320 500 g Poly Pail
18601321 2.5 kg Poly Pail
18601322 12 kg Poly Pail
18601323 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601320 Supplier Spectrum Chemical Mfg Cor Supplier No. CA150500GM
Only null left

CA150, 1305-62-0, Ca(OH)2

Spectrum™ Chemical Calcium Hydroxide, Powder, USP, EP, BP, JP is used as an astringent that is topically applied in solutions or lotions. Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Color White
CAS Max % 100%
Percent Purity 95 to 100.5%
Grade Multi-Compendial
Packaging Poly Pail
Quantity 500 g
Chemical Name or Material Calcium Hydroxide