missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, Anhydrous, powder, ≥97%, Honeywell Fluka™
Anhydrous, powder,≥97%
Supplier: Honeywell Chemicals C49011KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Calcium chloride | |
| 1 kg | |
| CaCl2 | |
| NONH for all modes of transport | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| calcium;dichloride | |
| 5284359 | |
| 110.98g/mol | |
| Poly bottle |
| 10043-52-4 | |
| CaCl2 | |
| MFCD00010903 | |
| calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
| [Cl-].[Cl-].[Ca++] | |
| 110.98 | |
| CHEBI:3312 | |
| ≥97% | |
| 0.01 mmHg (20°C) |
Chemical Identifiers
| 10043-52-4 | |
| 110.98 | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| 5284359 | |
| [Cl-].[Cl-].[Ca++] |
| CaCl2 | |
| MFCD00010903 | |
| calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
| CHEBI:3312 |
Safety and Handling
P305 + P351 + P338
RTECSNumber : EV9800000