missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Anhydrous, powder, ≥97%, Honeywell Fluka™

Catalog No. p-7121611
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047906 500 g Poly Bottle
6003026 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60047906 Supplier Honeywell Chemicals Supplier No. C4901500G
Only null left

Anhydrous, powder,≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point 1,900°C
Melting Point 772°C (literature)
CAS Max % 100.0000%
Physical Form Powder
Quantity 500 g
Linear Formula CaCl2
UN Number NONH for all modes of transport
IUPAC Name calcium dichloride
Formula Weight 110.98
Percent Purity >97%
Packaging Poly Bottle
Vapor Pressure 0.01 mmHg (20°C)
Density 2.150 g/cm3
Chemical Name or Material Calcium chloride, Anhydrous, powder, ≥97%
Show More Show Less
Hazard Statement P305 + P351 + P338
ShelfLife 1620 days from date of manufacture
EINECSNumber 233-140-8
RTECSNumber EV9800000
Recommended Storage Room Temp