missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Anhydrous, Desiccant, 4 Mesh, Approx. 95%, Spectrum™ Chemical

C1075, 10043-52-4, CaCl2

Supplier:  Spectrum Chemical Mfg Cor C107525KGCS4

 View more versions of this product

Catalog No. 18601241


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Calcium Chloride, Anhydrous, Desiccant, 4 Mesh is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and must be kept in tightly sealed, air tight containers. Calcium chloride has many uses but its main use is as a desiccant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Specifications

Calcium Chloride, Anhydrous, Desiccant
100%
CaCl2
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
∼95%
10043-52-4
4 x 2.5 kg
Poly Pail
[Cl-].[Cl-].[Ca++]
110.98
Reagent

Chemical Identifiers

10043-52-4
110.98
calcium dichloride
CaCl2
UXVMQQNJUSDDNG-UHFFFAOYSA-L
[Cl-].[Cl-].[Ca++]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions