missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate Standard , (1mL = 1mg CaCO3 = 0.4mg Ca), for Hardness, Certified, 1.000 ±0.010mg CaCO3/mL, LabChem™

Calcium Carbonate Standard, CCaO3, CAS Number-471-34-1, 7647-01-0, 500mL, 98.9, 1, 0.1, CHEBI:3311, Colorless, 1.000 +/- 0.010mg CaCO3/mL, 1g/mL, 100.09, Certified, Warning, GHS H Statement Causes skin irritation. Causes serious eye irritation.

$54.63 - $84.97

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC127007
View Documents
LabChem
LC127007
125 mL Poly Bottle
Each for $54.63
Only null left
 
LC127001
View Documents
LabChem
LC127001
500 mL Poly Bottle
Each for $84.97
Only null left
 
Description

Description

Calcium Carbonate Standard, CCaO3, CAS Number-471-34-1, 7647-01-0, 500mL, 98.9, 1, 0.1, CHEBI:3311, Colorless, 1.000 +/- 0.010mg CaCO3/mL, 1g/mL, 100.09, Certified, Warning, GHS H Statement

Causes skin irritation.

Causes serious eye irritation.

Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
10112
calcium carbonate

Specifications

Poly Bottle
98.9,1,0.1
1g/mL
CCaO3
CaCO3
Colorless
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
125 mL
CHEBI:3311
Certified
471-34-1,7647-01-0,7732-18-5
Passes Test
Liquid
1.000 ±0.010mg CaCO3/mL
MFCD00010906
Soluble in water
[Ca++].[O-]C([O-])=O
100.09
10112
100.09
Calcium Carbonate Standard, (1mL = 1mg CaCO3 = 0.4mg Ca), for Hardness
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at

Warning

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'