Learn More
Calcium Carbonate Standard , (1mL = 1mg CaCO3 = 0.4mg Ca), for Hardness, Certified, 1.000 ±0.010mg CaCO3/mL, LabChem™
Calcium Carbonate Standard, CCaO3, CAS Number-471-34-1, 7647-01-0, 500mL, 98.9, 1, 0.1, CHEBI:3311, Colorless, 1.000 +/- 0.010mg CaCO3/mL, 1g/mL, 100.09, Certified, Warning, GHS H Statement Causes skin irritation. Causes serious eye irritation.
Supplier: LabChem LC127001
Description
Calcium Carbonate Standard, CCaO3, CAS Number-471-34-1, 7647-01-0, 500mL, 98.9, 1, 0.1, CHEBI:3311, Colorless, 1.000 +/- 0.010mg CaCO3/mL, 1g/mL, 100.09, Certified, Warning, GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Specifications
| Calcium Carbonate Standard | |
| Poly Bottle | |
| 98.9,1,0.1 | |
| 1g/mL | |
| CCaO3 | |
| CaCO3 | |
| Colorless | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| calcium carbonate | |
| 500 mL | |
| CHEBI:3311 | |
| Certified |
| (1mL = 1mg CaCO3 = 0.4mg Ca), for Hardness | |
| 471-34-1,7647-01-0,471-34-1 | |
| Passes Test | |
| Liquid | |
| 1.000 ±0.010mg CaCO3/mL | |
| MFCD00010906 | |
| Soluble in water | |
| [Ca++].[O-]C([O-])=O | |
| 100.09 | |
| 10112 | |
| 100.09 |
Chemical Identifiers
| 471-34-1 | |
| 100.09 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| CHEBI:3311 | |
| [Ca++].[O-]C([O-])=O |
| CCaO3 | |
| MFCD00010906 | |
| 10112 | |
| calcium carbonate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Warning
Recommended Storage : Room Temperature