missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium acetate hydrate, Puriss., meets analytical spec. of FCC, E263, 99.0-100.5% (based on anhydrous substance), Honeywell Fluka™
Puriss., Meets analytical specification of FCC, E263, 99.0-100.5% (based on anhydrous substance)
$642.47 - $642.47
Chemical Identifiers
| CAS | 114460-21-8 |
|---|---|
| Molecular Formula | C4H6CaO4 |
| Molecular Weight (g/mol) | 158.17 |
| MDL Number | MFCD00012448 |
| InChI Key | VSGNNIFQASZAOI-UHFFFAOYSA-L |
| IUPAC Name | calcium diacetate |
| SMILES | [Ca++].CC([O-])=O.CC([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 114460-21-8 | |
| 158.17 | |
| VSGNNIFQASZAOI-UHFFFAOYSA-L | |
| [Ca++].CC([O-])=O.CC([O-])=O |
| C4H6CaO4 | |
| MFCD00012448 | |
| calcium diacetate |
Specifications
| 114460-21-8 | |
| C4H6CaO4 | |
| MFCD00012448 | |
| 3692527 | |
| [Ca++].CC([O-])=O.CC([O-])=O | |
| 158.17 | |
| 99 to 100.5% (based on anhydrous substance) | |
| Calcium Acetate Hydrate, Meets analytical specification of FCC |
| 1 kg | |
| (CH3COO)2Ca · xH2O | |
| NONH for all modes of transport | |
| VSGNNIFQASZAOI-UHFFFAOYSA-L | |
| calcium diacetate | |
| 158.17g/mol (anhydrous basis) | |
| puriss. |