missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, Puriss., meets analytical spec. of FCC, E263, 99.0-100.5% (based on anhydrous substance), Solstice

Catalog No. 6002221
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002221 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002221 Supplier Solstice Supplier No. 250111KG

Puriss., Meets analytical specification of FCC, E263, 99.0-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 1 kg
Linear Formula (CH3COO)2Ca · xH2O
UN Number NONH for all modes of transport
Beilstein 3692527
Formula Weight 158.17g/mol (anhydrous basis)
Percent Purity 99 to 100.5% (based on anhydrous substance)
Grade puriss.
Chemical Name or Material Calcium Acetate Hydrate, Meets analytical specification of FCC