missing translation for 'onlineSavingsMsg'
Learn More

Calcium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Ca), LabChem™

Catalog No. p-5388647
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC126507 125 mL Poly Bottle
LC126501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC126507 Supplier LabChem Supplier No. LC126507
Only null left

Chemical Identifiers

CAS 471-34-1 , 7697-37-2 , 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 98.75, 1, 0.25
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Ca)
Linear Formula CaCO3
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 100.09
Grade Certified
Chemical Name or Material Calcium AA Standard, 1000ppm (1mL = 1mg Ca)
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature