missing translation for 'onlineSavingsMsg'
Learn More

Calcium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Ca), LabChem™

Supplier:  LabChem LC126501

 View more versions of this product

Catalog No. LC126501


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Calcium AA Standard
Poly Bottle
98.75,1,0.25
1g/mL
CCaO3
CaCO3
Colorless
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
500 mL
CHEBI:3311
Certified
1000ppm (1mL = 1mg Ca)
471-34-1,7697-37-2,471-34-1
Passes Test
Liquid
1000 ppm ±10 ppm (1mL = 1mg Ca)
MFCD00010906
Soluble in water
[Ca++].[O-]C([O-])=O
100.09
10112
100.09

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
10112
calcium carbonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions