missing translation for 'onlineSavingsMsg'
Learn More

Calcium AA Standard, 1000 ppm Ca in 3% HNO3, Ricca Chemical

$113.47 - $360.93

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
ACA1KN-100
View Documents
Ricca Chemical Company
ACA1KN100
100 mL Natural LDPE Bottle
Each for $113.47
Only null left
 
ACA1KN-500
SDS View Documents
Ricca Chemical Company
ACA1KN500
500 mL Natural Poly Bottle
Each for $360.93
Only null left
 
Description

Description

Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
10112
calcium carbonate

Specifications

Natural LDPE Bottle
1,000ppm Ca
97.0,4.99,0.25
Liquid
MFCD00010906
< 2
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
100 mL
CHEBI:3311
471-34-1 , 7697-37-2 , 471-34-1
93.0,4.8,0.24
1 mL = 1 mg Ca
CCaO3
Colorless
Approximately 0°C
[Ca++].[O-]C([O-])=O
100.09
10112
Calcium AA Standard
Videos
SDS
Documents

Documents