missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Troponin I, Human Heart

Catalog No. 648480100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
648480100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 648480100UG Supplier MilliporeSigma™ Supplier No. 648480100UG

Purified from human heart.

EMD Millipore Calbiochem™ Troponin I, Human Heart is used as a sty ard in immunoassay, immunogen for antisera production.

  • Inhibitory subunit, binds to actin in absence of Ca2+, inhibits ATPase activity of actomyosin
  • Presence of hcTnI in plasma, used for early detection of myocardial infarction
  • Useful as tracer for iodination
  • Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg for antibodies to HIV HCV
  • Lyophilized from 10mM HCl
  • Assay: ≥95% by SDS-PAGE
  • CAS: 77108-40-8

Specifications

Quantity 100 μg
Content And Storage -20°C