missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ SMCC, Water-Soluble

Catalog No. 57311550MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57311550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57311550MG Supplier MilliporeSigma™ Supplier No. 57311550MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for coupling enzymes to antibodies.

EMD Millipore Calbiochem™ SMCC, Water-Soluble contains cyclohexane bridge, which gives extra stability to maleimide group.

  • Enzyme activity, antibody specificity are preserved after coupling
  • Assay: ≥94% by HPLC
  • CAS: 92921-24-9
  • Molecular Formula: C16H17N2O9S · Na
  • Packaging: Glass Bottle
  • Color: Off-White

Recommended Storage

2° to 8°C

Specifications

Quantity 50 mg
Product Type SMCC, heterobifunctional
Synonym Sulfosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulfo-SMCC
Content And Storage 2°C to 8°C