missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Sialyl Lewis x, Sodium Salt

Catalog No. 5659501MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5659501MG 1 mg
5659505MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5659501MG Supplier MilliporeSigma™ Supplier No. 5659501MG

Acts as minimal binding lig for E-selectins.

EMD Millipore Calbiochem™ Sialyl Lewis x, Sodium Salt is an enzymatically prepared tetrasaccharide.

  • Reduces injury, ischemia, reperfusion, immune complex-induced acute lung injury in animal models
  • Blocks E-selectin-mediated neutrophil recruitment to sites of inflammation in animal models
  • Assay: ≥95% by TLC
  • CAS: 98603-84-0
  • Molecular Formula: C31H51NaN2O23
  • Color: White

Specifications

Quantity 1 mg
Content And Storage -20°C