missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ RNase A, Bovine Pancreas

Catalog No. 5567450KU
Change view
Click to view available options
Quantity:
50 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5567450KU 50 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5567450KU Supplier MilliporeSigma™ Supplier No. 5567450KU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For selective degradation of RNA

  • Chromatographically purified
  • pyrimidine-specific endoribonuclease that acts on single-stranded RNA

Specifications

Content And Storage -20°C
Source Bovine pancreas
Quantity 50 kU
Product Type Ribonuclease A
Form Lyophilized

For Research use Only