missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Ribonucleic Acid, Yeast

Catalog No. 55714250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55714250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55714250MG Supplier MilliporeSigma™ Supplier No. 55714250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Life Science reasearch application

tRNA from Baker's yeast, prepared by the method of Holley, et al.

Specifications

CAS 63231-63-0
Color White to off-white
Content And Storage Store at −20°C
Quantity 250 mg
Form Solid