missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Polyinosinic Acid · Polycytidylic Acid, Sodium Salt, Double-Stranded

Catalog No. 52890610MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52890610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52890610MG Supplier MilliporeSigma™ Supplier No. 52890610MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Known to induce interferon production

  • Hypochromicity: ≥40% at 248nm 
  • Extinction coefficient = 5000 M-1cm-1 at 260nm

Specifications

CAS 42424-50-0
Color White
Content And Storage Store at +2°C to +8°C
pH 7.0
Purity ≥99% by TLC
Quantity 10 mg
Form Powder