Learn More
MilliporeSigma™ Calbiochem™ PhosphoDetect™ Phosphotyrosine Detection Kit
Provides set of four different monoclonal antibodies specific for tyrosine phosphorylation sites.
Supplier: MilliporeSigma™ 5253221KIT
Description
EMD Millipore Calbiochem™ PhosphoDetect™ Phosphotyrosine Detection Kit reacts with tyrosine phosphorylated proteins.
- Recognition is dependent on phosphorylation the surrounding amino acid motif
- Reacts with tyrosine phosphorylated proteins in chicken, human, mouse, rat, Xenopus, yeast, zebrafish. Phosphorylation patterns in cell extracts may differ when probed with different antibodies
- Packaging: Glass Bottle
Recommended Storage
−20°C
Specifications
| PhosphoDetect Phosphotyrosine Detection Kit | |
| 25μg of ea. of the phosphotyrosine antibodies including clones 2C8, 3B12, 9H8, and 16F4, and data sheet | |
| 1 Kit |