Learn More
MilliporeSigma™ Calbiochem™ PhosphoDetect™ Phosphothreonine Detection Kit
Provides set of three different monoclonal antibodies specific for threonine phosphorylation sites.
Supplier: MilliporeSigma™ 5252881KIT
Description
EMD Millipore Calbiochem™ PhosphoDetect™ Phosphothreonine Detection Kit reacts with threonine phosphorylated proteins.
- Recognition is dependent on phosphorylation the surrounding amino acid motif
- Reacts with threonine phosphorylated proteins in Arabidopsis, bacteria, chicken, human, maize, mouse, rat, tobacco, Xenopus, yeast, zebrafish
- Phosphorylation patterns in cell extracts may differ when probed with different antibodies
- Packaging: Glass Bottle
Recommended Storage
−20°C
Specifications
| PhosphoDetect Phosphothreonine Detection Kit | |
| 25μg of ea. of the Phosphothreonine Antibodies including clones 1E11, 4D11, and 14B3, Rabbit Muscle Positive Control, and data sheet | |
| 1 Kit |