missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ PBS 10X Liquid Concentrate, Molecular Biology Grade

Catalog No. 650420L
Change view
Click to view available options
Quantity:
4L
20L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
650420L 20L
65054L 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 650420L Supplier MilliporeSigma™ Supplier No. 650420L
Only null left

Makes 10L of 1X PBS Buffer from each liter.

  • Molecular Biology Grade
  • DNase: None Detected
  • pH (10%, water) at 25°C: 7.3 to 7.5
  • RNase: None Detected
  • Sterility: Passes test

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color Clear, Colorless
Quantity 20L