missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ DNA Extraction Buffer

Catalog No. 370020L
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
370020L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 370020L Supplier MilliporeSigma™ Supplier No. 370020L

Used as extraction buffer with pH ranging from 6.9 to 7.1.

  • Specifications:
  • Conductivity at 25°C: 35 to 55mS
  • DNase: None Detected
  • pH at 25°C: 6.9 to 7.1
  • RNase: None Detected
  • Titration (molarity): 225 to 260mM
  • Color: Clear colorless

Specifications

Content And Storage 15°C to 30°C
Quantity 20 L