missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Melittin

Catalog No. 444605250UG
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
444605250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 444605250UG Supplier MilliporeSigma™ Supplier No. 444605250UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits adenylate cyclase activity in synaptic membranes

  • 26-residue polypeptide from bee venom that binds calmodulin in a Ca2+-dependent manner
  • Used for affinity purification of several Calcium-binding proteins

Specifications

Content And Storage -20°C
Quantity 250 μg
Product Type Melittin
Form Lyophilized

For Research use Only